Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218995

Sigma-Aldrich

N,N-Dimethyl-1-naphthylamine

99%

Synonym(s):

1-Dimethylaminonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7N(CH3)2
CAS Number:
Molecular Weight:
171.24
Beilstein/REAXYS Number:
1424075
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

assay

99%

refractive index

n20/D 1.622 (lit.)

bp

139-140 °C/13 mmHg (lit.)

density

1.042 g/mL at 25 °C (lit.)

SMILES string

CN(C)c1cccc2ccccc12

InChI

1S/C12H13N/c1-13(2)12-9-5-7-10-6-3-4-8-11(10)12/h3-9H,1-2H3

InChI key

AJUXDFHPVZQOGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Ingham et al.
Zentralblatt fur Bakteriologie : international journal of medical microbiology, 279(2), 225-230 (1993-06-01)
A rapid swab method using either a dry or moist swab soaked with potassium nitrate/benzalkonium chloride solution, and a method employing a single powdered reagent were assessed as possible alternatives to the conventional test for the detection of bacterial nitrate
[Sensitivity of methods for determining nitrites].
P A Kal'tianis et al.
Laboratornoe delo, (3)(3), 27-28 (1982-01-01)
Chiharu Mizuguchi et al.
Biochimica et biophysica acta, 1841(12), 1716-1724 (2014-10-05)
Human apolipoprotein E (apoE) isoforms exhibit different conformational stabilities and lipid-binding properties that give rise to altered cholesterol metabolism among the isoforms. Using Trp-substituted mutations and site- directed fluorescence labeling, we made a comprehensive comparison of the conformational organization of
Jonas Camillus Jeppesen et al.
Langmuir : the ACS journal of surfaces and colloids, 31(46), 12699-12707 (2015-10-27)
Gel domains in lipid bilayers are structurally more complex than fluid domains. Growth dynamics can lead to noncircular domains with a heterogeneous orientational texture. Most model membrane studies involving gel domain morphology and lateral organization assume the domains to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service