Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

201529

Sigma-Aldrich

N-(4-Amino-2,5-diethoxyphenyl)benzamide

Dye content 90 %

Synonym(s):

4′-Amino-2′,5′-diethoxybenzanilide, Azoic Diazo No. 20, Fast blue BB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O3
CAS Number:
Molecular Weight:
300.35
Colour Index Number:
37175
Beilstein/REAXYS Number:
3416889
MDL number:
UNSPSC Code:
41116121
PubChem Substance ID:
Pricing and availability is not currently available.

form

powder

composition

Dye content, 90%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CCOc1cc(NC(=O)c2ccccc2)c(OCC)cc1N

InChI

1S/C17H20N2O3/c1-3-21-15-11-14(16(22-4-2)10-13(15)18)19-17(20)12-8-6-5-7-9-12/h5-11H,3-4,18H2,1-2H3,(H,19,20)

InChI key

CNXZLZNEIYFZGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service