Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197998

Sigma-Aldrich

Methyl 4,5-dimethoxy-2-nitrobenzoate

97%

Synonym(s):

Methyl 6-nitroveratrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H2(OCH3)2CO2CH3
CAS Number:
Molecular Weight:
241.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

141-144 °C (lit.)

SMILES string

COC(=O)c1cc(OC)c(OC)cc1[N+]([O-])=O

InChI

1S/C10H11NO6/c1-15-8-4-6(10(12)17-3)7(11(13)14)5-9(8)16-2/h4-5H,1-3H3

InChI key

SYYKLKHBZGFKOC-UHFFFAOYSA-N

Application

Methyl 4,5-dimethoxy-2-nitrobenzoate was used in the synthesis of methyl 4,5-dimethoxy-2-aminobenzoate[1].

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Reusable Polymer-anchored Palladium Catalyst for Reduction of Nitroorganics, Alkenes, Alkynes and Schiff Bases.
Saha C.
J. Chem. Res. Synop., (1), 44-45 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service