Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197114

Sigma-Aldrich

(4-Chloro-2-methylphenoxy)acetic acid sodium salt

≥95%

Synonym(s):

MCPA sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8ClNaO3
CAS Number:
Molecular Weight:
222.60
Beilstein/REAXYS Number:
5190432
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

≥95%

SMILES string

[Na+].Cc1cc(Cl)ccc1OCC([O-])=O

Pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harald Cederlund et al.
Journal of environmental management, 191, 28-34 (2017-01-14)
We studied the ability of a wood-based biochar to reduce the leaching of the pesticides chlorpyrifos, diuron, glyphosate and MCPA in a sand column test system. In addition, time-dependent adsorption of the pesticides to the biochar and to the sand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service