Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197068

Sigma-Aldrich

2,6-Dibromo-4-methylaniline

99%

Synonym(s):

2,6-Dibromo-p-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H2(CH3)NH2
CAS Number:
Molecular Weight:
264.95
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

mp

74-76 °C (lit.)

SMILES string

Cc1cc(Br)c(N)c(Br)c1

InChI

1S/C7H7Br2N/c1-4-2-5(8)7(10)6(9)3-4/h2-3H,10H2,1H3

InChI key

ATDIROHVRVQMRO-UHFFFAOYSA-N

General description

2,6-Dibromo-4-methylaniline undergoes Suzuki-coupling reaction with 2-dihydroxyboryl-3-methyl-2-cyclopenten-1-one during synthesis of the dinuclear dichlorotitanium complexes[1].

Application

2,6-Dibromo-4-methylaniline was used as coupling reagent in spectrophotometric determination of carbaryl in various environmental samples[2]. It was also in preparation of 2-bromo-6-iodo-4-methylaniline[3].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of reagents for the sensitive spectrophotometric determination of carbaryl in environmental samples.
Suresh KK, et al.
Analytical Letters, 38(4), 697-709 (2005)
Sang Hoon Lee et al.
Dalton transactions (Cambridge, England : 2003), (40)(40), 4608-4614 (2007-10-12)
Bimetallic dichlorotitanium complexes, {2,6-[eta(5)-2,5-Me2C5H2](2)-4-R-C6H2N-microN}{Ti(IV)Cl2}2 (, R=Me; , R=F) and 4,4'-A[{2-(eta(5)-2,3,5-Me3C5H)C6H3NC6H11-kappaN}Ti(IV)Cl2]2 (, A=CH2; , A=O; , A=ortho-C6H4) are prepared via a key step of the Suzuki-coupling reaction of 2-dihydroxyboryl-3-methyl-2-cyclopenten-1-one () with dibromo-compounds. The solid state structure of was determined by X-ray
Asymmetric reactions of axially chiral amides: use of removable ortho-substituents in radical cyclizations of o-iodoacrylanilides and N-allyl-N-o-iodoacrylamides.
Petit M, et al.
Tetrahedron, 60(35), 7543-7552 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service