Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

195901

Sigma-Aldrich

3-(3,4-Dichlorophenoxy)benzaldehyde

94%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OC6H4CHO
CAS Number:
Molecular Weight:
267.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

94%

form

solid

refractive index

n20/D 1.619 (lit.)

bp

153 °C/0.2 mmHg (lit.)

density

1.348 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(Oc2cccc(C=O)c2)cc1Cl

InChI

1S/C13H8Cl2O2/c14-12-5-4-11(7-13(12)15)17-10-3-1-2-9(6-10)8-16/h1-8H

InChI key

ABQHJSHFFLAGHF-UHFFFAOYSA-N

General description

3-(3,4-Dichlorophenoxy)benzaldehyde on condensation with acetophenone yields chalcone[1].

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Automated parallel synthesis of chalcone-based screening libraries.
Powers DG, et al.
Tetrahedron, 54(16), 4085-4096 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service