Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

194581

Sigma-Aldrich

5,5-Dimethyl-1-pyrroline N-oxide

97%

Synonym(s):

DMPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein/REAXYS Number:
107603
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.496 (lit.)

bp

75 °C/0.4 mmHg (lit.)

mp

25-29 °C (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

CC1(C)CCC=[N+]1[O-]

InChI

1S/C6H11NO/c1-6(2)4-3-5-7(6)8/h5H,3-4H2,1-2H3

InChI key

VCUVETGKTILCLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Neuroprotective agent; nitric oxide spin trap. Used to study radicals formed by enzymatic acetaldehyde oxidation.
Neuroprotective agent; nitric oxide spin trap. Used to study radicals formed by enzymatic acetaldehyde oxidation. Incubation of lymphocytes with DMPO decreased DNA damage by NiCl2.
Spin-trapping reagent used to study the formation of hydroxyl[1][2] and superoxide[2] radicals.

replaced by

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

203.0 °F - closed cup

flash_point_c

95 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Pou et al.
Analytical biochemistry, 217(1), 76-83 (1994-02-15)
The spin trap 5,5-dimethyl-1-pyrroline-1-oxide (DMPO) alone, as well as DMPO or N-tert-butyl-alpha-phenylnitrone (PBN) in the presence of excess dimethyl sulfoxide (Me2SO), have been used as spin trapping systems for the detection of hydroxyl radical. However, the instability of DMPO and
Free Radical Res. Commun., 17, 377-377 (1992)
Sambuddha Banerjee et al.
Free radical biology & medicine, 53(6), 1317-1326 (2012-07-31)
We compared oxygenation and anaerobic oxidation reactions of a purified complex of human hemoglobin (Hb) and haptoglobin (Hb-Hp) to those of uncomplexed Hb. Under equilibrium conditions, Hb-Hp exhibited active-site heterogeneity and noncooperative, high-affinity O(2) binding (n(1/2)=0.88, P(1/2)=0.33 mm Hg in
Xiaoguang Duan et al.
Small (Weinheim an der Bergstrasse, Germany), 11(25), 3036-3044 (2015-03-20)
Sulfur and nitrogen co-doped reduced graphene oxide (rGO) is synthesized by a facile method and demonstrated remarkably enhanced activities in metal-free activation of peroxymonosulfate (PMS) for catalytic oxidation of phenol. Based on first-order kinetic model, S-N co-doped rGO (SNG) presents
Jun Di et al.
Nanoscale, 7(26), 11433-11443 (2015-06-18)
Novel carbon quantum dot (CQD) modified Bi2MoO6 photocatalysts were prepared via a facile hydrothermal process. The CQD modified Bi2MoO6 materials were characterized by multiple techniques. The CQDs with the average size of about 7 nm were distributed on the surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service