About This Item
SMILES string
CCC1(O)[C@@H]2CCC[C@H]1CCC2
InChI
1S/C11H20O/c1-2-11(12)9-5-3-6-10(11)8-4-7-9/h9-10,12H,2-8H2,1H3/t9-,10+,11?
InChI key
COQRKPRIDRDOOG-ZACCUICWSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service