Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

191744

Sigma-Aldrich

3-Methyl-1-phenyl-2-phospholene 1-oxide

85%, technical grade

Synonym(s):

1-Oxo-3-methyl-1-phenyl-2-phospholene, 2,3-Dihydro-4-methyl-1-phenylphosphole 1-oxide, MPPO, NSC 107634

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13OP
CAS Number:
Molecular Weight:
192.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

assay

85%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

impurities

5% dichloromethane

refractive index

n20/D 1.5707 (lit.)

bp

150 °C/0.15 mmHg (lit.)

SMILES string

CC1=CP(=O)(CC1)c2ccccc2

InChI

1S/C11H13OP/c1-10-7-8-13(12,9-10)11-5-3-2-4-6-11/h2-6,9H,7-8H2,1H3

Inchi Key

YMKWWHFRGALXLE-UHFFFAOYSA-N

Application

Catalyst for:
  • Intramolecular aza-Wittig cyclization reactions
  • Polymerization reactions

Reactant for preparation of:
  • Phospha sugars and their radical bromination derivatives as anticancer agents

Pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phuong-Mai Nguyen et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(10), 1703-1720 (2017-04-05)
Migration due to indirect contact with packaging caused several major sanitary crises, including the spread contamination of dry food by mineral oils and printing ink constituents from cardboard. The issues are still not fully resolved because the mechanisms have been
Andrea Walzl et al.
Molecules (Basel, Switzerland), 25(15) (2020-08-06)
The current demand to cut back on the use of plastic materials has brought a major boost to the search for bio-based alternatives. Not only are plastic bags and primary food packaging under scrutiny here, but also those materials used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service