189111
Sodium diethyldihydroaluminate solution
25 wt. % in toluene
Synonym(s):
OMH-1, Sodium aluminum diethyl dihydride
About This Item
Recommended Products
concentration
25 wt. % in toluene
impurities
5% tetrahydrofuran
SMILES string
[Na+].CC[AlH2-]CC
InChI
1S/2C2H5.Al.Na.2H/c2*1-2;;;;/h2*1H2,2H3;;;;/q;;-1;+1;;
InChI key
DJNBMRGHGJSTBH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Legal Information
signalword
Danger
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service