Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

188824

Sigma-Aldrich

2-Biphenylmethanol

99%

Synonym(s):

2-Phenylbenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH2OH
CAS Number:
Molecular Weight:
184.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

solid

bp

96 °C/0.04 mmHg (lit.)

mp

46-48 °C (lit.)

SMILES string

OCc1ccccc1-c2ccccc2

InChI

1S/C13H12O/c14-10-12-8-4-5-9-13(12)11-6-2-1-3-7-11/h1-9,14H,10H2

InChI key

VKTQADPEPIVMHK-UHFFFAOYSA-N

General description

2-Biphenylmethanol exhibits the formation of a new phase in a supercooled liquid state which was studied by differential scanning calorimetry and x-ray diffraction measurements[1]. On heating 2-biphenylmethanol undergoes transformation from the glassy to liquid phase which has been investigated by luminescence spectra[2].

Application

2-Biphenylmethanol was used in the synthesis of o-ethynylbiphenyl and o-alkynylstyrene derivatives[3].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annulation selectivity in the coupling of Fischer carbene complexes with o-alkynylbiphenyl and o-alkynylstyrene derivatives.
Jackson TJ and Herndon JW.
Tetrahedron, 57(18), 3859-3868 (2001)
Experimental evidence of formation of a new phase in supercooled liquid 2-biphenylmethanol.
Baran J, et al.
Journal of Physics. Condensed Matter : An Institute of Physics Journal, 18(24), 5695-5695 (2006)
Metastable polymorph formation in a supercooled liquid 2-biphenylmethanol: Luminescence study.
Baran J, et al.
Journal of Molecular Liquids, 153(2), 174-177 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service