Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

184780

Sigma-Aldrich

2-Phenethylbenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2C6H4CH2OH
CAS Number:
Molecular Weight:
212.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

form

solid

mp

56-59 °C (lit.)

SMILES string

OCc1ccccc1CCc2ccccc2

InChI

1S/C15H16O/c16-12-15-9-5-4-8-14(15)11-10-13-6-2-1-3-7-13/h1-9,16H,10-12H2

InChI key

CPZRYQJPVUJHOS-UHFFFAOYSA-N

General description

2-Phenethylbenzyl alcohol reacts with phosphorous tribromide to yield 2-phenethylbenzyl bromide[1].

Application

2-Phenethylbenzyl alcohol was used to prepare reagent required during the synthesis of non-symmetric cyclic sulfamide HIV-1 protease inhibitors[1].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a small library of non-symmetric cyclic sulfamide HIV-1 protease inhibitors.
Ax A, et al.
Tetrahedron, 66(23), 4049-4056 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service