Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

164186

Sigma-Aldrich

7-Methoxy-2-tetralone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
Beilstein/REAXYS Number:
1953198
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

refractive index

n20/D 1.566 (lit.)

bp

124-126 °C/1.5 mmHg (lit.)

density

1.13 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc2CCC(=O)Cc2c1

InChI

1S/C11H12O2/c1-13-11-5-3-8-2-4-10(12)6-9(8)7-11/h3,5,7H,2,4,6H2,1H3

InChI key

XEAPZXNZOJGVCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

7-Methoxy-2-tetralone was used in the synthesis of 2-bromo-3,4-dihydro-7-methoxy-1-naphthaldehyde[1] and 2-substituted octahydrobenzo[f]quinolines[2].

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dajie Li et al.
Bioorganic & medicinal chemistry, 11(17), 3795-3805 (2003-08-07)
Substituted benzo[i]phenanthridines that have incorporated within their structure an 8,9-methylenedioxy group can exhibit topoisomerase I-targeting activity. Structure-activity studies were performed to examine the influence of saturation at the 11,12-positions of several substituted 8,9-methylenedioxybenzo[i]phenanthridines. The activities of these dihydro analogues were
J C Craig et al.
Journal of medicinal chemistry, 32(5), 961-968 (1989-05-01)
A series of 2-substituted octahydrobenzo[f]quinolines has been synthesized and assayed for dopamine agonist activity. Only the compounds corresponding to the beta-rotameric conformation of dopamine showed biphasic activity in competition binding studies with the radioligand [3H]spiroperidol. These findings suggest that the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service