Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

163481

Sigma-Aldrich

2-Methoxy-5-nitrobenzyl bromide

97%

Synonym(s):

2-Bromomethyl-4-nitroanisole, Koshland’s Reagent II

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)CH2Br
CAS Number:
Molecular Weight:
246.06
Beilstein/REAXYS Number:
1878596
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

76-78 °C (lit.)

SMILES string

COc1ccc(cc1CBr)[N+]([O-])=O

InChI

1S/C8H8BrNO3/c1-13-8-3-2-7(10(11)12)4-6(8)5-9/h2-4H,5H2,1H3

InChI key

JRHMPHMGOGMNDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxy-5-nitrobenzyl bromide is a potential mechanism-based inactivator of enzymes that perform O-dealkylations[1].

Application

2-Methoxy-5-nitrobenzyl bromide was used to modify tryptophan residues of mouse interferon to enhance their activity[2]. It was also used as reagent for sulfhydryl modification[3].

Biochem/physiol Actions

2-Methoxy-5-nitrobenzyl bromide inactivates cytochrome P-450 in male rat liver microsomes[1].

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rusckowski et al.
Journal of interferon research, 2(2), 177-185 (1982-01-01)
Mouse interferon (MuIFN) preparations containing both the alpha and beta species were treated with chemical modifying reagents specific for lysine, cysteine and tryptophan residues. The purpose was to ascertain whether reagents which modify specific amino acid residues distinguished MuIFN-alpha from
A P Armstrong et al.
Drug metabolism and disposition: the biological fate of chemicals, 27(6), 741-745 (1999-05-29)
Mechanism-based inactivators serve as probes of enzyme mechanism, function, and structure. Koshland's Reagent II (2-methoxy-5-nitrobenzyl bromide, KR-II) is a potential mechanism-based inactivator of enzymes that perform O-dealkylations. The major phenobarbital-inducible form of cytochrome P-450 in male rat liver microsomes, CYP2B1
Aldrichimica Acta, 18, 78-78 (1985)
[Comparison of auc values for caffeine and its metabolites after single and repeated administration of 2-methoxy-5-nitrobenzyl bromide].
Jakub Bojakowski et al.
Roczniki Panstwowego Zakladu Higieny, 54 Suppl, 56-56 (2003-07-30)
I V Kosarev et al.
Biochimica et biophysica acta, 790(2), 125-131 (1984-10-23)
The effect of the chemical modification of lysine, histidine, arginine, tyrosine, tryptophan residues and carboxylic groups on the cryoproperties of monoclonal human cryoglobulin M has been studied. The modification of 35-40 lysine residues and that of 42-45 arginine residues in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service