Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

163279

2,2′,3,3′,4,4′,5,5′,6,6′-PCB

99%

Synonym(s):

Decachlorobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12Cl10
CAS Number:
Molecular Weight:
498.66
Pricing and availability is not currently available.

assay

99%

InChI

1S/C12Cl10/c13-3-1(4(14)8(18)11(21)7(3)17)2-5(15)9(19)12(22)10(20)6(2)16

InChI key

ONXPZLFXDMAPRO-UHFFFAOYSA-N

Application

Reference standard for the analysis of PCB′s.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ovokeroye A Abafe et al.
Environmental science and pollution research international, 22(18), 14078-14086 (2015-05-12)
Workplace exposure to persistent organic pollutants is a concern for human health. This study examined the presence of polybrominated diphenyl ethers (PBDEs) and polychlorinated biphenyls (PCBs) in the indoor dust from two major e-waste recycling sites and a university electronic
Photodegradation of decachlorobiphenyl.
M Koshioka et al.
Bulletin of environmental contamination and toxicology, 38(3), 409-415 (1987-03-01)
A morphologic and physiologic bioassay for quantifying alterations in the lung following experimental chronic inhalation of tobacco smoke.
G L Huber et al.
Bulletin europeen de physiopathologie respiratoire, 17(2), 269-327 (1981-01-01)
Perchlorination reaction applied to the rapid determination of PCBs.
K Takamiya
Bulletin of environmental contamination and toxicology, 30(5), 600-605 (1983-05-01)
Xing Han et al.
Toxicology, 255(3), 177-186 (2008-11-22)
2,2',3,3',4,4',5,5',6,6'-Decachlorobiphenyl (PCB 209) is a fully chlorinated, non-coplanar biphenyl. To demonstrate that PCB 209 is not likely to exhibit human health hazards common to coplanar PCBs it was tested for cytochrome P450 (P450) enzyme induction potentials, genetic toxicity, and endocrine-modulating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service