Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

161411

Sigma-Aldrich

α-Chlorocinnamaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C(Cl)CHO
CAS Number:
Molecular Weight:
166.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.64 (lit.)

bp

107-108 °C/3 mmHg (lit.)

mp

20-22 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl\C(C=O)=C/c1ccccc1

InChI

1S/C9H7ClO/c10-9(7-11)6-8-4-2-1-3-5-8/h1-7H/b9-6-

InChI key

SARRRAKOHPKFBW-TWGQIWQCSA-N

General description

α-Chlorocinnamaldehyde inhibits the corrosion of ARMCO iron in 4 M HCl[1]. It undergoes chemoselective reduction in the presence of calcium amidoborane to yield allylic alcohol[2].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron corrosion inhibition in hot 4 M HCl solution by t-cinnamaldehyde and its structure-related compounds.
Zucchi F, et al.
Corrosion Science, 36(10), 1683-1690 (1994)
Calcium amidoborane, a new reagent for chemoselective reduction of a, ?-unsaturated aldehydes and ketones to allylic alcohols.
Xu W, et al.
Royal Society of Chemistry Advances, 2(14), 6005-6010 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service