Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

156310

Sigma-Aldrich

Methacryloyl chloride

technical grade, 90%

Synonym(s):

2-Methyl-2-propenoyl chloride, 2-Methylpropenoic acid chloride, Methacrylyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COCl
CAS Number:
Molecular Weight:
104.53
Beilstein/REAXYS Number:
878175
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

technical grade

assay

90%

contains

200 ppm phenothiazine as stabilizer

refractive index

n20/D 1.442 (lit.)

bp

95-96 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(Cl)=O

InChI

1S/C4H5ClO/c1-3(2)4(5)6/h1H2,2H3

InChI key

VHRYZQNGTZXDNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

55.0 °F - closed cup

flash_point_c

12.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Asmussen et al.
Scandinavian journal of dental research, 91(2), 153-155 (1983-04-01)
The bonding of restorative resins to dentin by means of an intermediary monomer containing a carboxylic acid chloride or an isocyanate group was investigated. Such intermediary monomers are supposed to react with the organic constituent of dentin. Bond strengths were
Xiangzheng Hu et al.
Steroids, 70(8), 531-537 (2005-05-17)
Bile acids in the family of steroid compounds can be chemically modified for biochemical and other applications. Derivatives of cholic acid with multiple methacrylate groups can be prepared by the use of methacrylic acid, methacryloyl chloride and methacryloyl anhydride as
Polymerized phosphatidyl choline vesicles. Stabilized and controllable time-release carriers.
S L Regen et al.
Biochemical and biophysical research communications, 101(1), 131-136 (1981-07-16)
Huai-Song Wang et al.
Journal of chromatography. A, 1409, 132-137 (2015-07-23)
Novel biocompatible chiral monolithic stationary phase was prepared by reverse and direct atom transfer radical polymerization (ATRP) methods. By taking advantages of the controlled/living property of ATRP method, the chiral monolith was prepared by reverse ATRP (RATRP) firstly. An attractive
Petra Votavova et al.
Journal of biomedical nanotechnology, 11(9), 1662-1673 (2015-10-22)
Interleukin-2 (IL-2) possesses a strong stimulatory activity for activated T and NK cells and it is an attractive molecule for immunotherapy. Nevertheless, extremely short half-life and severe toxicities associated with high-dose IL-2 treatment are serious and limiting drawbacks. In order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service