Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

155985

Sigma-Aldrich

Methyl 4-nitrobenzoate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CO2CH3
CAS Number:
Molecular Weight:
181.15
Beilstein/REAXYS Number:
1912198
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

≥99%

mp

94-96 °C (lit.)

SMILES string

COC(=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C8H7NO4/c1-13-8(10)6-2-4-7(5-3-6)9(11)12/h2-5H,1H3

InChI key

YOJAHJGBFDPSDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service