Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15466

Sigma-Aldrich

Boc-Aib-OH

≥99.0% (T)

Synonym(s):

α-(Boc-amino)isobutyric acid, Boc-α-methylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CNHC(CH3)2CO2H
CAS Number:
Molecular Weight:
203.24
Beilstein/REAXYS Number:
1953772
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

Quality Level

assay

≥99.0% (T)

form

solid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

118-122 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)NC(C)(C)C(O)=O

InChI

1S/C9H17NO4/c1-8(2,3)14-7(13)10-9(4,5)6(11)12/h1-5H3,(H,10,13)(H,11,12)

InChI key

MFNXWZGIFWJHMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Protected unusual and seldom-occurring achiral amino acid.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Mayr et al.
Liebigs Ann. Chem., 715-715 (1980)
John R Horsley et al.
Journal of the American Chemical Society, 136(35), 12479-12488 (2014-08-15)
Electrochemical studies are reported on a series of peptides constrained into either a 310-helix (1-6) or β-strand (7-9) conformation, with variable numbers of electron rich alkene containing side chains. Peptides (1 and 2) and (7 and 8) are further constrained

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service