Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15410

Sigma-Aldrich

Boc-Tyr(Bzl)-OH

≥99.0% (T)

Synonym(s):

Boc-O-benzyl-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4CH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
371.43
Beilstein/REAXYS Number:
2227416
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

assay

≥99.0% (T)

optical activity

[α]20/D +27.0±1.5°, c = 2% in ethanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

110-112 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1ccc(OCc2ccccc2)cc1)C(O)=O

InChI

1S/C21H25NO5/c1-21(2,3)27-20(25)22-18(19(23)24)13-15-9-11-17(12-10-15)26-14-16-7-5-4-6-8-16/h4-12,18H,13-14H2,1-3H3,(H,22,25)(H,23,24)/t18-/m0/s1

InChI key

ZAVSPTOJKOFMTA-SFHVURJKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan Z Dreger et al.
Biomaterials, 221, 119399-119399 (2019-08-20)
Hernia repair outcomes have improved with more robust material options for surgeons and optimized surgical techniques. However, ventral hernia repairs remain challenging with an inherent risk of post-surgical adhesions in the peritoneal space which can occur regardless of interventional material
Karel Sedlar et al.
Genomics, 113(1 Pt 2), 1109-1119 (2020-11-10)
Clostridium diolis DSM 15410 is a type strain of solventogenic clostridium capable of conducting isopropanol-butanol-ethanol fermentation. By studying its growth on different carbohydrates, we verified its ability to utilize glycerol and produce 1,3-propanediol and discovered its ability to produced isopropanol.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service