152013
2,3-Dimethyl-4-methoxybenzaldehyde
99%
Synonym(s):
2,3-Dimethylanisaldehyde
About This Item
Recommended Products
assay
99%
form
solid
mp
52-55 °C (lit.)
functional group
aldehyde
SMILES string
[H]C(=O)c1ccc(OC)c(C)c1C
InChI
1S/C10H12O2/c1-7-8(2)10(12-3)5-4-9(7)6-11/h4-6H,1-3H3
InChI key
MAFDJCNDRCNZFM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service