Skip to Content
MilliporeSigma
All Photos(1)

Documents

15067

Sigma-Aldrich

Boc-D-Asp(OBzl)-OH

≥98.0% (HPLC)

Synonym(s):

Boc-D-aspartic acid 4-benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2O2CCH2CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
323.34
Beilstein/REAXYS Number:
2305471
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (HPLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@H](CC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C16H21NO6/c1-16(2,3)23-15(21)17-12(14(19)20)9-13(18)22-10-11-7-5-4-6-8-11/h4-8,12H,9-10H2,1-3H3,(H,17,21)(H,19,20)/t12-/m1/s1

InChI key

SOHLZANWVLCPHK-GFCCVEGCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service