Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

147397

Sigma-Aldrich

2,4-Dichloro-5-sulfamoylbenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(SO2NH2)CO2H
CAS Number:
Molecular Weight:
270.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

230-232 °C (dec.) (lit.)

SMILES string

NS(=O)(=O)c1cc(C(O)=O)c(Cl)cc1Cl

InChI

1S/C7H5Cl2NO4S/c8-4-2-5(9)6(15(10,13)14)1-3(4)7(11)12/h1-2H,(H,11,12)(H2,10,13,14)

Inchi Key

ZSHHRBYVHTVRFK-UHFFFAOYSA-N

Gene Information

human ... CA2(760)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ohnishi et al.
Antimicrobial agents and chemotherapy, 22(2), 250-254 (1982-08-01)
Sodium 5-aminosulfonyl-2,4-dichlorobenzoate (M12325) was evaluated for antiviral activity in tissue culture and infected mice. At concentrations ranging from 2.5 to 75.8 micrograms/ml, M12325 inhibited the cytopathic effects of 10 mean tissue culture infective doses of influenza virus A/WSN, A/FM, A/Kumamoto

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service