Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

142425

Sigma-Aldrich

2-(4-Pyridyl)ethanesulfonic acid

97%

Synonym(s):

4-Pyridineethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO3S
CAS Number:
Molecular Weight:
187.22
Beilstein/REAXYS Number:
136690
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

297-299 °C (dec.) (lit.)

SMILES string

OS(=O)(=O)CCc1ccncc1

InChI

1S/C7H9NO3S/c9-12(10,11)6-3-7-1-4-8-5-2-7/h1-2,4-5H,3,6H2,(H,9,10,11)

InChI key

RGIIAYDCZSXHGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(4-Pyridyl)ethanesulfonic acid is the sulfonated substrate for alkanesulfonate monooxygenase isolated from Escherichia coli[1].

application

2-(4-Pyridyl)ethanesulfonic acid was used in the preparation of benzyldimethyltetradecyl ammoniumchloride dihydrate solution[2].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Eichhorn et al.
The Journal of biological chemistry, 274(38), 26639-26646 (1999-09-10)
The Escherichia coli ssuEADCB gene cluster is required for the utilization of alkanesulfonates as sulfur sources, and is expressed under conditions of sulfate or cysteine starvation. The SsuD and SsuE proteins were overexpressed and characterized. SsuE was purified to homogeneity
On-site Analysis of Trace Inorganic Components in Environ-mental Water after Preconcentration with Membrane Filter Col-lection and by Tristimulus Colorimetry and Spectrophotometry.
KASAHARA I, et al.
Analytical Sciences, 17, i1233-i1233 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service