Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

140260

Sigma-Aldrich

3-Amino-1,2,4-triazole-5-thiol

95%

Synonym(s):

3-Amino-5-mercapto-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4N4S
CAS Number:
Molecular Weight:
116.14
Beilstein/REAXYS Number:
112404
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

mp

>300 °C (lit.)

solubility

water: soluble 25 mg/mL, clear, faintly yellow to yellow

SMILES string

Nc1n[nH]c(S)n1

InChI

1S/C2H4N4S/c3-1-4-2(7)6-5-1/h(H4,3,4,5,6,7)

InChI key

WZUUZPAYWFIBDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Amino-1,2,4-triazole-5-thiol inhibits corrosion of copper in aerated acidic chloride pickling solutions[1].

Application

3-Amino-1,2,4-triazole-5-thiol was used to study the inhibition of corrosion of iron in 3.5% NaCl solutions by low concentrations of ATT and 1,1′-thiocarbonyldiimidazole[2]. It was used to prepare surface-enhanced Raman scattering based pH nano- and microsensor using silver nanoparticles[3].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

El-Sayed M Sherif et al.
Journal of colloid and interface science, 306(1), 96-104 (2006-11-07)
Corrosion of copper in aerated acidic chloride pickling (0.5 M HCl) solutions and its inhibition by 3-amino-1,2,4-triazole-5-thiol (ATT) have been investigated using electrochemical techniques and weight-loss measurements, along with Raman spectroscopy. Electrochemical measurements for copper after varied immersion periods of
Corrosion Inhibition in Chloride Solutions of Iron by 3-Amino-1, 2, 4-triazole-5-thiol and 1, 1'-Thiocarbonyldiimidazole.
Sherif E-SM.
International Journal of Electrochemical Science, 7, 4834-4846 (2012)
Gözde Aydoğdu Tığ et al.
Talanta, 210, 120666-120666 (2020-01-29)
In this work, we reported an electrochemical aptasensor based on the poly-3-amino-1,2,4-triazole-5-thiol/graphene oxide composite (P(ATT)-GO) and gold nanoparticles (AuNPs) modified graphite screen-printed electrode (GSPE) (GSPE/P(ATT)-GO/AuNPs) for determination of lipocalin-2 (LCN2) (neutrophil gelatinase-associated lipocalin). A sandwich based strategy was utilized to
Srinu Nagireddi et al.
International journal of biological macromolecules, 127, 320-329 (2019-01-15)
This article targets the efficacy of chitosan cross-linked 3-Amino-1,2,4-triazole-5-thiol derivative for the recovery of Pd from synthetic electroless plating solutions (ELP) whose solution chemistry complexity is brought forward with ethylenediaminetetraacetic acid (EDTA), ammonium hydroxide (NH4OH), and surfactant (cetyl trimethyl ammonium
SERS-based pH sensor utilizing 3-amino-5-mercapto-1, 2, 4-triazole functionalized Ag nanoparticles.
Piotrowski P, et al.
Analyst (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service