Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

139491

Sigma-Aldrich

Methylglyoxal bis(guanylhydrazone) dihydrochloride hydrate

99%

Synonym(s):

Methyl GAG dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C[=NNHC(=NH)NH2]CH[=NNHC(=NH)NH2] · 2HCl · xH2O
CAS Number:
Molecular Weight:
257.12 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

mp

255 °C (dec.) (lit.)

SMILES string

O.Cl.CC(C=NNC(N)=N)=NNC(N)=N

InChI

1S/C5H12N8.ClH.H2O/c1-3(11-13-5(8)9)2-10-12-4(6)7;;/h2H,1H3,(H4,6,7,12)(H4,8,9,13);1H;1H2

InChI key

DHFAMLHAUITXFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of methylglyoxal-bis(guanylhydrazone) on polyamine metabolism in spleens of mice with disseminated L1210 lymphoid leukemia.
O Heby et al.
Cancer research, 34(4), 886-892 (1974-04-01)
Methyl glyoxal bis(guanylhydrazone) as a potent inhibitor of mammalian and yeast S-adenosylmethionine decarboxylases.
H G Williams-Ashman et al.
Biochemical and biophysical research communications, 46(1), 288-295 (1972-01-14)
Paradoxical enhancement of S-adenosylmethionine decarboxylase in rat tissues following administration of the specific inhibitor methyl glyoxal bis(guanylhydrazone).
A E Pegg et al.
Biochemical and biophysical research communications, 52(2), 696-701 (1973-05-15)
C W Porter et al.
Cancer research, 40(7), 2330-2335 (1980-07-01)
The use of methylglyoxal-bis(guanylhydrazone) (MGBG) in the clinical treatment of myeloid and lymphoid disorders has been limited by severe host toxicity to renewing tissues, particularly the intestinal mucosa. Since the drug is a potent inhibitor of spermidine biosynthesis, the distributions
Polyamines and polyamine-metabolizing enzyme activities in human semen.
J Jänne et al.
Clinica chimica acta; international journal of clinical chemistry, 48(4), 393-401 (1973-11-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service