Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

137405

1,1,1-Trichloro-2-propanol

99%

Synonym(s):

1,1,1-Trichloroisopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5Cl3O
CAS Number:
Molecular Weight:
163.43
PubChem Substance ID:
Pricing and availability is not currently available.

assay

99%

InChI

1S/C3H5Cl3O/c1-2(7)3(4,5)6/h2,7H,1H3

InChI key

HCMBPASAOZIEDZ-UHFFFAOYSA-N

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service