Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

135852

Sigma-Aldrich

4,4′-Methylenebis(cyclohexylamine)

97%

Synonym(s):

4,4′-Diaminodicyclohexylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(C6H10NH2)2
CAS Number:
Molecular Weight:
210.36
Beilstein/REAXYS Number:
2079179
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor pressure

<0.1 mmHg ( 38 °C)

assay

97%

density

0.95 g/mL at 25 °C (lit.)

SMILES string

NC1CCC(CC1)CC2CCC(N)CC2

InChI

1S/C13H26N2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h10-13H,1-9,14-15H2

InChI key

DZIHTWJGPDVSGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

318.2 °F - closed cup

flash_point_c

159 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G L Kennedy
Journal of applied toxicology : JAT, 11(5), 367-371 (1991-10-11)
1,4-Bis(aminocyclohexyl)methane (PACM; CAS No. 1761-71-3) is a white solid considered for use in polymeric systems. PACM is moderately toxic systemically, with the acute oral lethal dose in rats between 670 and 1000 mg kg-1. PACM is a weak dermal sensitizer
Malin Frick et al.
Contact dermatitis, 48(6), 305-309 (2003-10-09)
From August 1999 to April 2001, there was an outbreak of severe eczema at a factory manufacturing medical equipment. A glue, mainly based on the isocyanate dicyclohexylmethane-4,4'-diisocyanate (DMDI), was suspected as being the cause of the problem. 16 workers with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service