129941
1-Methyl-3-nitro-1-nitrosoguanidine
97%
Synonym(s):
N-Methyl-N′-nitro-N-nitrosoguanidine, Diazomethane precursor, MNNG
About This Item
Recommended Products
assay
97%
mp
118 °C (dec.) (lit.)
SMILES string
CN(N=O)C(=N)N[N+]([O-])=O
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
Other Notes
comparable product
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2
Storage Class
4.1A - Other explosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service