Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

126985

Sigma-Aldrich

N,N-Dimethyl-N′-ethylethylenediamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NHCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
116.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.4222 (lit.)

bp

134-135 °C (lit.)

density

0.738 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCNCCN(C)C

InChI

1S/C6H16N2/c1-4-7-5-6-8(2)3/h7H,4-6H2,1-3H3

InChI key

WLNSKTSWPYTNLY-UHFFFAOYSA-N

General description

N,N-Dimethyl-N′-ethylethylenediamine is prepared by the reaction of NiX2 • 6 H2O (X = NO3-, ClO4-), bidentate amine, [Cu(oxpn)] [oxpn = N,N′-bis(3-aminopropyl)oxamide] and sodium nitrite in ethanolic solution[1].

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

75.2 °F - closed cup

flash_point_c

24 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reactions Between Dinuclear CuII?NiII Complexes and the Nitrito Ligand: A New One-Dimensional Complex [Cu(oxpn)Ni(?-NO2)(trimen)]n(ClO4)n with Nitrite Acting as a Tridentate Bridging Ligand and Ferro- and Antiferromagnetic Alternation.
Diaz C, et al.
European Journal of Inorganic Chemistry, 2000(4), 675-681 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service