All Photos(1)
About This Item
Linear Formula:
CH3(CH2)3NHCH2CN
CAS Number:
Molecular Weight:
112.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
assay
97%
refractive index
n20/D 1.434 (lit.)
bp
85 °C/9 mmHg (lit.)
density
0.891 g/mL at 25 °C (lit.)
SMILES string
CCCCNCC#N
InChI
1S/C6H12N2/c1-2-3-5-8-6-4-7/h8H,2-3,5-6H2,1H3
InChI key
SPMMZKAWNAKPJM-UHFFFAOYSA-N
General description
(Butylamino)acetonitrile may be used in preparation of amides, carbamates and sulphonamides.
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service