Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

117889

Sigma-Aldrich

2-Chloro-5-nitrobenzophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)COC6H5
CAS Number:
Molecular Weight:
261.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

83-85 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Cl)c(c1)C(=O)c2ccccc2

InChI

1S/C13H8ClNO3/c14-12-7-6-10(15(17)18)8-11(12)13(16)9-4-2-1-3-5-9/h1-8H

InChI key

HRPHZUAPQWJPCZ-UHFFFAOYSA-N

Application

2-Chloro-5-nitrobenzophenone was used in the synthesis of sulfone-activated difluoro monomers with benzoyl substituents: bis(2-benzoyl-4-fluorophenyl) sulfone and 2-benzoyl-4,4′-difluorophenyl sulfone[1].It was used in derivatization of phenols during the synthesis of 13C- and 14C-uniformly labeled catechol[2].

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of aromatic poly (ether sulfone) s with pendent benzoyl groups.
Eom HJ and Kim SY.
Polym. Bull., 41(6), 631-637 (1998)
Synthesis of 13C-and 14C-labelled catechol.
Ji R and Schaffer A.
Journal of Labelled Compounds & Radiopharmaceuticals, 45(7), 551-558 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service