About This Item
assay
97%
mp
53-56 °C (lit.)
SMILES string
Cc1c(Cl)ccc(c1Cl)[N+]([O-])=O
InChI
1S/C7H5Cl2NO2/c1-4-5(8)2-3-6(7(4)9)10(11)12/h2-3H,1H3
InChI key
WBNZUUIFTPNYRN-UHFFFAOYSA-N
General description
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service