Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04537

Sigma-Aldrich

Diisopropyl(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane

≥95%

Synonym(s):

(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl)diisopropylsilane, Diisopropyl(1H,1H,2H,2H-perfluorodecyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19F17Si
CAS Number:
Molecular Weight:
562.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

form

liquid

refractive index

n20/D 1.357

density

1.385 g/mL at 25 °C

SMILES string

[H][Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(C(C)C)C(C)C

InChI

1S/C16H19F17Si/c1-7(2)34(8(3)4)6-5-9(17,18)10(19,20)11(21,22)12(23,24)13(25,26)14(27,28)15(29,30)16(31,32)33/h7-8,34H,5-6H2,1-4H3

InChI key

BFRDVGYAJGMBJI-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

target_organs

Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service