Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADVH97EBCE62

AChemBlock

2-(1-iodoethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
281.93
MDL number:
Product ADVH97EBCE62 is currently not available for purchase. Contact Technical Service

Iupac Name

2-(1-iodoethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Inchi Code

1S/C8H16BIO2/c1-6(10)9-11-7(2,3)8(4,5)12-9/h6H,1-5H3

InChI key

WAGSGVLOYSWUSL-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

0.97

Country of Origin

CN

Physical Form

light yellow liquid

Product Link

https://www.achemblock.com/u134968-2-1-iodoethyl-4-4-5-5-tetramethyl-1-3-2-dioxaborolane.html

Synonym

2-(1-iodoethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/u134968-2-1-iodoethyl-4-4-5-5-tetramethyl-1-3-2-dioxaborolane.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library