Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-975

AAT Bioquest

AAT Bioquest Cyanine 5 azide

25 mg

Synonym(s):

Cy5® azide

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$764.00

$764.00


Available to ship onApril 23, 2025Details



Select a Size

Change View
1 EA
$764.00

About This Item

Molecular Weight:
838.91
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?

$764.00


Available to ship onApril 23, 2025Details


fluorescence

λex 651 nm;λem 670 nm

manufacturer/tradename

AAT Bioquest

General description

A variety of cyanine 5 ( Cy5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis. Our Cy5® Fluors are thoroughly QC tested to ensure high levels of chromophore and reactive dye content. Mono-reactive dyes are suitable for targeted, precise labeling of proteins and oligonucleotides and bis-reactive dyes are more suitable for general labeling. NHS ester dyes are recommended for labeling amine groups and maleimide dyes are recommended for labeling thiol groups. This Cy5® azide selectively reacts with alkyne groups.

Cyanine 5 azide [equivalent to Cy5® azide] is manufactured by AAT Bioquest

Legal Information

Cy5® is a trademark of GE Healthcare

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library