Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-75

AAT Bioquest

AAT Bioquest Rhodamine 101 Chloride Salt

25 mg

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$90.00

$90.00


Available to ship onMarch 11, 2025Details



Select a Size

Change View
1 EA
$90.00

About This Item

CAS Number:
Molecular Weight:
527.05
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?

$90.00


Available to ship onMarch 11, 2025Details


fluorescence

λex 569 nm;λem 590 nm

manufacturer/tradename

AAT Bioquest

General description

Rhodamine dyes are used extensively in a variety of biological applications such as fluorescence microscopy, flow cytometry, fluorescence correlation spectroscopy and ELISA.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library