Skip to Content
MilliporeSigma
All Photos(1)

Documents

AATB-22211

AAT Bioquest

AAT Bioquest Rhodamine B, hexyl ester, perchlorate

10 mg

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
627.17
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?

fluorescence

λex 546 nm;λem 567 nm

manufacturer/tradename

AAT Bioquest

General description

Positively charged rhodamine dyes (such as rhodamine esters and rosamines) are selectively localized in mitochondria, thus they are widely used for labeling mitochondria of live cells.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library