Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-21064

AAT Bioquest

AAT Bioquest Rhod-2, AM

20x50 ug UltraPure Grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
1123.96
Product AATB-21064 is currently not available for purchase. Contact Technical Service

fluorescence

λex 553 nm;λem 577 nm

manufacturer/tradename

AAT Bioquest

General description

The long-wavelength Rhod-2 Ca2+ indicators are valuable alternatives to Fluo-3 for experiments in cells and tissues that have high levels of autofluorescence. Rhod-2 AM is cell-permeable version of Rhod-2.

Rhod-2, AM *UltraPure Grade* *CAS#: 145037-81-6* is manufactured by AAT Bioquest

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library