Key Documents
AATB-21053
AAT Bioquest
AAT Bioquest BTC, tetrapotassium salt
1 mg
About This Item
CAS Number:
Product AATB-21053 is currently not available for purchase. Contact Technical Service
manufacturer/tradename
AAT Bioquest
General description
This cell-impermeant coumarin Ca2+ indicator exhibits a shift in excitation maximum from about 480 nm to 400 nm upon binding Ca2+, enabling ratiometric calcium measurements.
BTC, tetrapotassium salt *CAS 216453-54-2* is manufactured by AAT Bioquest
Choose from one of the most recent versions:
Certificates of Analysis (COA)