Key Documents
AATB-20590
AAT Bioquest
AAT Bioquest Cal Red™ R525/650 AM
1 mg
About This Item
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
AAT Bioquest
General description
Cal Red™ R525/650 has been developed as a new 488 nm-excitable ratiometric fluorescence calcium indicator. Cal Red™ R525/650 is well excited at 488 nm with two emissions at 525 nm and 650 nm. Upon binding calcium, the emission signal of Cal Red™ R525/650 is increased at 525 nm and decreased at 650 nm when excited at 488 nm. The excitation and emission wavelength of Cal Red™ R525/650 are compatible with common filter sets with minimal damage to cells, making it a robust tool for evaluating and screening GPCR agonists and antagonists as well as calcium channel targets.
Legal Information
Cal Red™ is a trademark of AAT Bioquest
Choose from one of the most recent versions:
Certificates of Analysis (COA)