Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-20507

AAT Bioquest

AAT Bioquest OG488 BAPTA-1, AM

10x50 ug Cell-permeant

Synonym(s):

Oregon Green® 488 BAPTA-1, AM

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
1258.07
Product AATB-20507 is currently not available for purchase. Contact Technical Service

fluorescence

λex 493 nm;λem 522 nm

manufacturer/tradename

AAT Bioquest

General description

OG488 BAPTA -1 is the same molecule of Oregon Green 488 BAPTA-1. It is a visible light-excitable calcium indicator that is often used with FITC filter set. OG 488 BAPTA-1 has excitation and emission at 494 and 523 nm respectively. Its fluorescence intensity increases by ~14 fold upon binding calcium ion.

OG488 BAPTA-1, AM [equivalent to Oregon Green® 488 BAPTA-1, AM] *Cell permeant* is manufactured by AAT Bioquest

Legal Information

OG488® is a trademark of AAT Bioquest, Oregon Green® is the Trademark of Invitrogen

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library