Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-20380

AAT Bioquest

AAT Bioquest Mag-Fura-2 tetrapotassium salt

1 mg Cell impermeant

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
586.68
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?

fluorescence

λex 336 nm;λem 505 nm

manufacturer/tradename

AAT Bioquest

General description

Mag-Fura-2, tetrapotassium salt is an intracellular magnesium indicator that is ratiometric and UV light-excitable. This water-soluble salt form is useful for intracellular loading by microinjection or infusion from patch pipette. This particular magnesium indicator is also used for high concentration calcium detection.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library