Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-179

AAT Bioquest

AAT Bioquest Cyanine 5.5 alkyne

1 mg

Synonym(s):

Cy5.5® alkyne

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
1068.14
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?

fluorescence

λex 683 nm;λem 703 nm

manufacturer/tradename

AAT Bioquest

General description

A variety of cyanine 5.5 (Cy5.5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.

Legal Information

Cy5.5® is a trademark of GE Healthcare

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library