Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-178

AAT Bioquest

AAT Bioquest Cyanine 5.5 azide

1 mg

Synonym(s):

Cy5.5® azide

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
1099.17
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?
Product AATB-178 is currently not available for purchase. Contact AAT Bioquest.

fluorescence

λex 683 nm;λem 703 nm

manufacturer/tradename

AAT Bioquest

General description

A variety of cyanine 5.5 (Cy5.5®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.

Cyanine 5.5 azide [equivalent to Cy5.5® azide] is manufactured by AAT Bioquest

Legal Information

Cy5.5® is a trademark of GE Healthcare

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library