Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-16051

AAT Bioquest

AAT Bioquest ROS Brite™ HPF

1 mg Optimized for Detecting Reactive Oxygen Species (ROS)

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
424.4
Product AATB-16051 is currently not available for purchase. Contact Technical Service

fluorescence

λex 498 nm;λem 517 nm

manufacturer/tradename

AAT Bioquest

General description

Reactive oxygen species (ROS) are chemically reactive molecules containing oxygen. ROS Brite™ HPF is a fluorogenic probe to measure hydroxyly radical in cells using conventional fluorescence microscopy, high-content imaging, microplate fluorometry, or flow cytometry. The cell-permeant ROS Brite™ HPF reagent is nonfluorescent and produces bright green fluorescence upon reaction with hydroxyl radical. The resulting fluorescence can be measured using fluorescence imaging, high-content imaging, microplate fluorometry, or flow cytometry. In the presence of peroxidase, HPF also reacts with hydrogen peroxide. HPF has good selectivity to hydroxyl radical compared to other ROS (e.g., supoeroxide and nitric oxide).

ROS Brite™ HPF *Optimized for Detecting Reactive Oxygen Species (ROS)* is manufactured by AAT Bioquest

Legal Information

ROS Brite™ is a trademark of AAT Bioquest

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library