Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-143

AAT Bioquest

AAT Bioquest Cyanine 3 azide

1 mg

Synonym(s):

Cy3® azide

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$115.00

$115.00


Available to ship onMay 06, 2025Details



Select a Size

Change View
1 EA
$115.00

About This Item

Molecular Weight:
812.88
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?

$115.00


Available to ship onMay 06, 2025Details


fluorescence

λex 555 nm;λem 569 nm

manufacturer/tradename

AAT Bioquest

General description

A variety of cyanine 3 (Cy3®) dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis. Cy3® dyes have enhanced fluorescence upon binding to proteins. Cy3® azide selectively reacts with alkyne groups.

Cyanine 3 azide [equivalent to Cy3® azide] is manufactured by AAT Bioquest

Legal Information

Cy3® is a trademark of GE Healthcare

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library