Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-13550

AAT Bioquest

AAT Bioquest Z-LRGG-AMC

1 mg

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$108.00

$108.00


Available to ship onApril 23, 2025Details



Select a Size

Change View
1 EA
$108.00

About This Item

Molecular Weight:
692.77
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?

$108.00


Available to ship onApril 23, 2025Details


fluorescence

λex 341 nm;λem 441 nm

manufacturer/tradename

AAT Bioquest

General description

Coronaviruses (CoVs) can infect humans and multiple species of animals, causing a wide spectrum of diseases. Papain-like protease (PLpro) of coronaviruses carries out proteolytic maturation of non-structural proteins that play a role in replication of the virus and performs deubiquitination of host cell factors to scuttle antiviral responses. Z-LRGG-AMC is cleaved by papain-like proteases to give the highly fluorescent AMC product. The fluorescence intensity of released AMC is proportional to the protease activity. Z-LRGG-AMC might be used for screening and studying kinetics of PLpro inhibitors.

Z-LRGG-AMC is manufactured by AAT Bioquest

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library