Documents
AATB-13413
AAT Bioquest
AAT Bioquest Z-IETD-pNA
5 mg
About This Item
CAS Number:
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
AAT Bioquest
General description
Z-IETD-pNA is a colorimetric caspase-8/granzyme B substrate containing the benzyloxycarbonyl (Z) moiety.