Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AATB-13411

AAT Bioquest

AAT Bioquest Ac-IETD-AMC

5 mg

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
675.68
PDF Icon
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
PDF Icon
Have a question?
Product AATB-13411 is currently not available for purchase. Contact AAT Bioquest.

fluorescence

λex 341 nm;λem 441 nm

manufacturer/tradename

AAT Bioquest

General description

Ac-IETD-AMC is a fluorogenic caspase-8/granzyme B substrate containing the acetyl (Ac) moiety.

Ac-IETD-AMC is manufactured by AAT Bioquest

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library